trans-Diethyl caronate 90%

Lieferant: Thermo Scientific

374260010
ACRO374260010EA 161 EUR
ACRO374260010
trans-Diethyl caronate 90%
trans-Diethyl caronate

Appearance: Clear colorless to light yellow Liquid

Formel: C₁₁H₁₈O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00145428
CAS-Nummer: 67945-22-6

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4420 to 1.4450 (20°C, 589 nm)
GC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR