3-Chlor-5-fluoranilin 97%

Lieferant: Thermo Fisher Scientific
Achtung

374740050
ACRO374740050EA 198 EUR
ACRO374740050
3-Chlor-5-fluoranilin 97%
3-Chlor-5-fluoranilin
Formel: C₆H₅ClFN
Molecular Weight: 145.56 g/mol
Dichte: 1.45 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD03407962
CAS-Nummer: 4863-91-6
UN: 2810
ADR: 6.1 III

Bestellen

Spezifikation Testergebnisse

Appearance Clear light yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.5610 to 1.5640 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR