N,N-Bis(trifluormethansulfonyl)anilin 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1,1,1-Trifluoro-N-phenyl-N-((trifluoromethyl)sulphonyl)methanesulphonamide, N-Phenylbis(trifluoromethanesulphonimide)
374790050
374790250
374791000
ACRO374790050EA
101
EUR
ACRO374790050
ACRO374790250
ACRO374791000
N,N-Bis(trifluormethansulfonyl)anilin 97%
N,N-Bis(trifluormethansulfonyl)anilin
Formel:
C₈H₅F₆NO₄S₂ Molecular Weight: 357.25 g/mol Schmelzpunkt: 99…105 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000404 CAS-Nummer: 37595-74-7 |
Spezifikation Testergebnisse
Appearance | White to yellow to beige Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 99 to 105 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...