(S)-(-)-3-Brom-α-methylbenzylalkohol 95%, 98% ee

Lieferant: Thermo Scientific
Achtung

Synonyme: (S)-1-(3-Bromophenyl)ethanol

376500010
ACRO376500010EA 149 EUR
ACRO376500010
(S)-(-)-3-Brom-α-methylbenzylalkohol 95%, 98% ee
(S)-(-)-3-Brom-α-methylbenzylalkohol
Formel: C₈H₉BrO
Molecular Weight: 201,06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06201859
CAS-Nummer: 134615-22-8

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥94 %
Enantiomeric excess ≥97.5 % (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR