Phenyl β-D-glucopyranoside hydrate 98%

Lieferant: Thermo Fisher Scientific

Synonyme: Phenyl-β-D-glucoside hydrate

376890050
ACRO376890050EA 98.4 EUR
ACRO376890050
Phenyl β-D-glucopyranoside hydrate 98%
Phenyl β-D-glucopyranoside hydrate

CAS No.:

Formel: C₁₂H₁₆O₆.xH₂O
Molecular Weight: 256.255 g/mol (anhy)
Schmelzpunkt: 172…176 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03410292
CAS-Nummer: 1266359-89-0

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Water ≤9 % (K.F.)
HPLC ≥97.5 %
Specific optical rotation (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR