N-(3-Hydroxypropyl)phthalimid 99%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-(3-Hydroxypropyl)-1,2-benzoldicarbonsäureimid, N-(3-Hydroxypropyl)phthalsäureimid
377010250
377011000
ACRO377010250EA
41.1
EUR
ACRO377010250
ACRO377011000
N-(3-Hydroxypropyl)phthalimid 99%
N-(3-Hydroxypropyl)phthalimid
Formel:
C₁₁H₁₁NO₃ Schmelzpunkt: 75…77 °C |
MDL:
MFCD00023097 CAS-Nummer: 883-44-3 EINECS: 212-931-1 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 75 to 78 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...