4-Methoxy-3-pyridinboronsäure Hydrat 97%

Lieferant: Thermo Fisher Scientific
Achtung

378382500 378380010
ACRO378382500EA 54.8 EUR
ACRO378382500 ACRO378380010
4-Methoxy-3-pyridinboronsäure Hydrat 97%
4-Methoxy-3-pyridinboronsäure Hydrat
Formel: C₆H₈BNO₃·H₂O
Lagertemperatur: Kühlschrank
MDL: MFCD07186213
CAS-Nummer: 1266615-79-5

Bestellen

Spezifikation Testergebnisse

Appearance Cream Powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR