1-Fluor-3-iod-5-nitrobenzol 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
378500010
ACRO378500010EA
74.3
EUR
ACRO378500010
1-Fluor-3-iod-5-nitrobenzol 98%
1-Fluoro-3-iodo-5-nitrobenzene
Formel:
C₆H₃FINO₂ Schmelzpunkt: 77…79 °C |
MDL:
MFCD00007219 CAS-Nummer: 3819-88-3 |
Spezifikation Testergebnisse
Appearance | Yellow to brown to khaki Powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 76 to 80 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...