Calciumbis(dihydrogenorthophosphat) 90%, rein

Lieferant: Thermo Scientific
Gefahr

Synonyme: Calcium phosphate, monobasic

378610010
ACRO378610010EA 75.8 EUR
ACRO378610010
Calciumbis(dihydrogenorthophosphat) 90%, rein
Calciumbis(dihydrogenorthophosphat)
Formel: H₄CaO₈P₂
Molecular Weight: 234,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00010898
CAS-Nummer: 7758-23-8
EINECS: 231-837-1

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Titration Complexometric ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR