2-Ethylbenzoesäure 97%

Lieferant: Thermo Fisher Scientific
Achtung

378750010 378750050
ACRO378750010EA 110 EUR
ACRO378750010 ACRO378750050
2-Ethylbenzoesäure 97%
2-Ethylbenzoic acid
Formel: C₉H₁₀O₂
Molecular Weight: 150.1700 g/mol
Schmelzpunkt: 62…66 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00045838
CAS-Nummer: 612-19-1

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 62 to 66 °C
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR