2-[N,N-Bis(trifluoromethanesulphonyl)amino]-5-chloropyridine
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-[N,N-Bis(Trifluoromethylsulphonyl)amino]-5-chloropyridine, N-(5-Chloro-2-pyridyl)triflimide, 2-[N,N-Bis(trifluoromethylsulphonyl)amino]-5-chloropyridine, N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulphonyl)imide, Comins Triflating Reagent, N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulphonimide)
378800050
378800250
ACRO378800050EA
138
EUR
ACRO378800050
ACRO378800250
2-[N,N-Bis(trifluoromethanesulphonyl)amino]-5-chloropyridine
2-[N,N-Bis(trifluoromethanesulphonyl)amino]-5-chloropyridine
Formel:
C₇H₃ClF₆N₂O₄S₂ Molecular Weight: 392.67 g/mol Siedepunkt: 77…90 °C (.2 mmHg) Schmelzpunkt: 45…47 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00191833 CAS-Nummer: 145100-51-2 |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystalline solid |
Infrared spectrum | Conforms |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...