2-Brom-5-chlorpyridin 98%

Lieferant: Thermo Fisher Scientific
Achtung

378980010 378980050
ACRO378980010EA 77.4 EUR
ACRO378980010 ACRO378980050
2-Brom-5-chlorpyridin 98%
2-Bromo-5-chloropyridine
Formel: C₅H₃BrClN
Molecular Weight: 192.44 g/mol
Schmelzpunkt: 65…69 °C
MDL: MFCD00234006
CAS-Nummer: 40473-01-6
UN: 2811

Bestellen

Spezifikation Testergebnisse

Appearance Beige to yellow-brown Powder
Infrared spectrum Conforms
Melting point 65 to 69 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR