Propargyl-p-toluolsulfonat 97%

Lieferant: Thermo Fisher Scientific
Achtung

379680050 379680250 379681000
ACRO379680050EA 38.8 EUR
ACRO379680050 ACRO379680250 ACRO379681000
Propargyl-p-toluolsulfonat 97%
Propargyl-p-toluolsulfonat
Formel: C₁₀H₁₀O₃S
Lagertemperatur: Kühlschrank
MDL: MFCD00078365
CAS-Nummer: 6165-76-0

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5270 to 1.5290 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR