Propargyl-p-toluolsulfonat 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
379680050
379680250
379681000
ACRO379680050EA
38.8
EUR
ACRO379680050
ACRO379680250
ACRO379681000
Propargyl-p-toluolsulfonat 97%
Propargyl-p-toluolsulfonat
Formel:
C₁₀H₁₀O₃S Lagertemperatur: Kühlschrank |
MDL:
MFCD00078365 CAS-Nummer: 6165-76-0 |
Spezifikation Testergebnisse
Appearance | Clear colorless to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5270 to 1.5290 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...