Methyltrifluorpyruvat 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Methyl 3,3,3-trifluoropyruvate, Methyl 3,3,3-trifluoro-2-oxopropanoate
379710050
ACRO379710050EA
60.7
EUR
ACRO379710050
Methyltrifluorpyruvat 97%
Methyltrifluorpyruvat
Formel:
C₄H₃F₃O₃ Molecular Weight: 156,06 g/mol Flammpunkt: 26 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00114936 CAS-Nummer: 13089-11-7 UN: 2924 ADR: 3,III |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3320 to 1.3340 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...