6-Brompicolinsäure 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
6-Bromo-2-pyridinecarboxylic acid, 6-Bromopyridine-2-carboxylic acid
379930010
ACRO379930010EA
39.2
EUR
ACRO379930010
6-Brompicolinsäure 98%
6-Brompicolinsäure
Formel:
C₆H₄BrNO₂ Molecular Weight: 202,01 g/mol Schmelzpunkt: 192…194 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00093197 CAS-Nummer: 21190-87-4 |
Spezifikation Testergebnisse
Appearance | Off-white to tan Powder |
Infrared spectrum | Conforms |
Titration with NaOH | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...