(4-Bromphenyl)aceton 97%

Lieferant: Thermo Fisher Scientific

380220050
ACRO380220050EA 65.4 EUR
ACRO380220050
(4-Bromphenyl)aceton 97%
(4-Bromphenyl)aceton
Formel: C₉H₉BrO
Lagertemperatur: Raumtemperatur
MDL: MFCD00210401
CAS-Nummer: 6186-22-7

Bestellen

Spezifikation Testergebnisse

Appearance Clear yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5560 to 1.5590 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR