Triformamide 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
trialkanoylamines, N,N-Diformylformamide, Formamide, N,N-diformyl-
380440050
ACRO380440050EA
96
EUR
ACRO380440050
Triformamide 97%
Triformamide
Formel:
C₃H₃NO₃ Molecular Weight: 101,06 g/mol Schmelzpunkt: 100…101 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08059356 CAS-Nummer: 25891-31-0 |
Spezifikation Testergebnisse
Appearance | White Crystals |
Infrared spectrum | Conforms |
Melting point | 95.0 to 100.0 °C |
Elemental Analysis | ≥96.0 % (ex C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...