3-Chlorphthalsäureanhydrid 95+%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Chloroisobenzofuran-1,3-dione

381290010 381290050 381290250
ACRO381290010EA 31.8 EUR
ACRO381290010 ACRO381290050 ACRO381290250
3-Chlorphthalsäureanhydrid 95+%
3-Chlorphthalsäureanhydrid
Formel: C₈H₃ClO₃
Molecular Weight: 182.56 g/mol
Schmelzpunkt: 123…126 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00023107
CAS-Nummer: 117-21-5
EINECS: 204-179-8

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder or flakes and/or chunks
Infrared spectrum Conforms
Melting point 122 to 126 °C
Titration by coupling ≥95.0 %

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR