Zellulose, mikrokristallin ∼ Ø 90 µm, reinst

Lieferant: Thermo Fisher Scientific

Synonyme: Zellulose

382312500 382310010 382310050 382310250
ACRO382312500EA 65.8 EUR
ACRO382312500 ACRO382310010 ACRO382310050 ACRO382310250
Zellulose, mikrokristallin ∼ Ø 90 µm, reinst
Zellulose
Formel: (C₁₂H₂₂O₁₁)n
Schmelzpunkt: 260 - 270 °C
Dichte: 1,27 to 1.6 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00081512
CAS-Nummer: 9004-34-6
EINECS: 232-674-9
Merck Index: 13,01977

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline or granular powder
Infrared spectrum Conforms
Loss on drying ≤1.5 %
Heavy metals ≤10 ppm
Particle size ≥70 % (> 32 µm)
Residue after ignition ≤0.05 %
pH 5 to 7


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR