Zellulose, mikrokristallin ∼ Ø 90 µm, reinst
Lieferant: Thermo Fisher Scientific
Synonyme:
Zellulose
382312500
382310010
382310050
382310250
ACRO382312500EA
65.8
EUR
ACRO382312500
ACRO382310010
ACRO382310050
ACRO382310250
Zellulose, mikrokristallin ∼ Ø 90 µm, reinst
Zellulose
Formel:
(C₁₂H₂₂O₁₁)n Schmelzpunkt: 260 - 270 °C Dichte: 1,27 to 1.6 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00081512 CAS-Nummer: 9004-34-6 EINECS: 232-674-9 Merck Index: 13,01977 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline or granular powder |
Infrared spectrum | Conforms |
Loss on drying | ≤1.5 % |
Heavy metals | ≤10 ppm |
Particle size | ≥70 % (> 32 µm) |
Residue after ignition | ≤0.05 % |
pH | 5 to 7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...