3-Amino-6-chlorpyridazin 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
6-Chlorpyridazin-3-amin
382830500
ACRO382830500EA
630
EUR
ACRO382830500
3-Amino-6-chlorpyridazin 97%
3-Amino-6-chlorpyridazin
Formel:
C₄H₄ClN₃ Molecular Weight: 129,55 g/mol Schmelzpunkt: 210…211 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051506 CAS-Nummer: 5469-69-2 |
Spezifikation Testergebnisse
Appearance | Off-white to light brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...