1H-Pyrazol-4-boronsäurepinakolester 97+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
382940010
382940050
382940250
ACRO382940010EA
85.5
EUR
ACRO382940010
ACRO382940050
ACRO382940250
1H-Pyrazol-4-boronsäurepinakolester 97+%
1H-Pyrazol-4-boronsäurepinakolester
Formel:
C₉H₁₅BN₂O₂ Schmelzpunkt: 125…130 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03453063 CAS-Nummer: 269410-08-4 |
Spezifikation Testergebnisse
Appearance | Off-white to tan Powder |
Infrared spectrum | Conforms |
GC | ≥97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...