Diglycidyl ether 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
383130050
383130010
ACRO383130050EA
215
EUR
ACRO383130050
ACRO383130010
Diglycidyl ether 95%
Diglycidyl ether
Appearance: Clear colorless to light brown Liquid
Formel:
C₆H₁₀O₃ Siedepunkt: 260 °C (760 mmHg) Dichte: 1,12 g/cm³ Flammpunkt: 64 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00080479 CAS-Nummer: 2238-07-5 UN: 2922 ADR: 8,III |
Spezifikation Testergebnisse
Appearance | Clear colorless to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4460 to 1.4500 (20°C, 589 nm) |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...