Chloroform 99.8% stabilisiert zur Analyse
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Stabilisierung:
Stabilisiert mit Amylen
383770010
383770025
383770250
ACRO383770010EA
63.6
EUR
ACRO383770010
ACRO383770025
ACRO383770250
Chloroform 99.8% stabilisiert zur Analyse
Chloroform
Formel:
CHCl₃ Siedepunkt: 61 °C (760 mmHg) Schmelzpunkt: –63 °C Dichte: 1,48 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000826 CAS-Nummer: 67-66-3 EINECS: 200-663-8 UN: 1888 ADR: 6.1,III REACH: 05-2114614906-45 Merck Index: 13,02160 |
|
Spezifikation Testergebnisse
Appearance | Clear liquid |
Water | ≤0.02 % (K.F.) |
GC | ≥99.8 % |
Color scale | ≤5 APHA |
Stabilizer | Approx. 100 ppm Amylene |
Residue after evaporation | ≤5 ppm |
Acidity/alkalinity | ≤0.0005 meq/g |
Suitability | (for use with Dithizone) Passes test |
Substance darkened by H2SO4 | ≤5 APHA |
Free chlorine | ≤5 ppm |
Calcium (Ca) | ≤0.1 ppm |
Copper (Cu) | ≤0.02 ppm |
Iron (Fe) | ≤0.1 ppm |
Lead (Pb) | ≤0.02 ppm |
Magnesium (Mg) | ≤0.05 ppm |
Potassium (K) | ≤0.1 ppm |
Sodium (Na) | ≤0.2 ppm |
Zinc (Zn) | ≤0.05 ppm |
Impurity | ≤0.005 % Trichloroethylene |
Total phosphorus (P) | ≤0.1 ppm |
Total silicon (Si) | ≤0.05 ppm |
Total sulfur (S) | ≤0.5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...