n-Hexan contains appr. 95% n-Hexane zur Analyse

Lieferant: Thermo Fisher Scientific
Gefahr

383800010 383800025 383800050 383800250
ACRO383800010EA 68.9 EUR
ACRO383800010 ACRO383800025 ACRO383800050 ACRO383800250
n-Hexan contains appr. 95% n-Hexane zur Analyse
n-Hexan
Formel: H₃C(CH₂)₄CH₃
Siedepunkt: 69 °C (760 mmHg)
Schmelzpunkt: –95 °C
Dichte: 0,659 g/cm³
Flammpunkt: –22 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009520
CAS-Nummer: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance Clear liquid
Water ≤0.02 % (K.F.)
GC ≥95.0 %
Color scale ≤10 APHA
Residue after evaporation ≤0.002 %
Acidity/alkalinity ≤0.00008 meq/g
Substance darkened by H2SO4 passes test
Calcium (Ca) ≤0.2 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.02 ppm
Magnesium (Mg) ≤0.05 ppm
Potassium (K) ≤0.1 ppm
Sodium (Na) ≤0.1 ppm
Zinc (Zn) ≤0.1 ppm
Hydrocarbons ≤0.1 % Aromatic hydrocarbons
Total phosphorus (P) ≤0.1 ppm
Total silicon (Si) ≤0.05 ppm
Total sulfur (S) ≤10 ppm
Bromine number ≤0.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR