Isohexane contains <5% n-Hexane for pesticide residue analysis

Lieferant: Thermo Fisher Scientific
Gefahr

383820010 383820025
ACRO383820010EA 63.7 EUR
ACRO383820010 ACRO383820025
Isohexane contains <5% n-Hexane for pesticide residue analysis
Isohexan (Isomerengemisch)
Formel: C₆H₁₄
Siedepunkt: 60 °C (760 mmHg)
Schmelzpunkt: –154 °C
Dichte: 0,66 g/cm³
Flammpunkt: –23 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179311
CAS-Nummer: 73513-42-5
EINECS: 295-570-2
UN: 1208
ADR: 3,II
REACH: 01-2119484651-34

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless Liquid
Water ≤0.01 % (Coulometric)
GC (ECD) Passes test, (NPD) Passes test, (FID) Passes test
Residue after evaporation ≤2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR