n-Pentan zur Analyse

Lieferant: Thermo Fisher Scientific
Gefahr

383900010 383900025
ACRO383900010EA 112 EUR
ACRO383900010 ACRO383900025
n-Pentan zur Analyse
n-Pentan
Formel: H₃C(CH₂)₃CH₃
Siedepunkt: 36 °C (760 mmHg)
Schmelzpunkt: –130 °C
Dichte: 0,626 g/cm³
Flammpunkt: –49 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009498
CAS-Nummer: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
REACH: 01-2119459286-30
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance Clear liquid
Water ≤0.01 % (K.F.)
GC ≥99.0 %
Color scale ≤10 APHA
Residue after evaporation ≤0.002 %
Acidity/alkalinity ≤0.0001 meq/g
Calcium (Ca) ≤0.5 ppm
Copper (Cu) ≤0.05 ppm
Iron (Fe) ≤0.2 ppm
Lead (Pb) ≤0.05 ppm
Magnesium (Mg) ≤0.1 ppm
Potassium (K) ≤0.2 ppm
Sodium (Na) ≤1 ppm
Zinc (Zn) ≤0.1 ppm
Total phosphorus (P) ≤0.5 ppm
Total silicon (Si) ≤0.02 ppm
Total sulfur (S) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR