Titan(IV)oxid, Aeroxide® P25

Lieferant: Thermo Fisher Scientific

384290500 384292500 384290010
ACRO384290500EA 67.8 EUR
ACRO384290500 ACRO384292500 ACRO384290010
Titan(IV)oxid, Aeroxide® P25
Titan(IV)oxid
Formel: TiO₂
Siedepunkt: 2900 °C (760 mmHg)
Schmelzpunkt: 1855 °C
Dichte: 4,23 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00011269
CAS-Nummer: 13463-67-7
EINECS: 236-675-5
Merck Index: 13,09549

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Assay ≥99.5 % (on ignited substance) Typical
Specific surface area 35 to 65 m²/g
pH 3.5 to 4.5 (4 % dispersion)
Acidity (HCl) ≤0.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR