4-Bromo-2-Fluorobenzylamine 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: (4-Bromo-2-fluorophenyl)methanamine

384440050
ACRO384440050EA 85.3 EUR
ACRO384440050
4-Bromo-2-Fluorobenzylamine 98%
4-Bromo-2-Fluorobenzylamine

Appearance: Clear colorless to light yellow Liquid

Formel: C₇H₇BrFN
Molecular Weight: 204,042 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00153076
CAS-Nummer: 112734-22-2
UN: 2735
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5630 to 1.5660 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR