1-Boc-1H-pyrazol-4-boronsäurepinakolester 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
384640010
384640050
ACRO384640010EA
73.6
EUR
ACRO384640010
ACRO384640050
1-Boc-1H-pyrazol-4-boronsäurepinakolester 97%
1-Boc-1H-pyrazol-4-boronsäurepinakolester
Formel:
C₁₄H₂₃N₂O₄B Schmelzpunkt: 82…89 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD05663873 CAS-Nummer: 552846-17-0 |
Spezifikation Testergebnisse
Appearance | White to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 82 to 89 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...