1-Boc-piperidin-4-carbonsäure 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-(tert-Butoxycarbonyl)piperidine-4-carboxylic acid, N-BOC-4-Piperidinecarboxylic acid, Boc-Inp-OH, 1-Boc-isonipecotic acid

384870010 384870050
ACRO384870010EA 29.4 EUR
ACRO384870010 ACRO384870050
1-Boc-piperidin-4-carbonsäure 98%
1-Boc-piperidin-4-carbonsäure
Formel: C₁₁H₁₉NO₄
Schmelzpunkt: 149…153 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00076999
CAS-Nummer: 84358-13-4

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 149 to 153 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR