Graphit, Pulver

Lieferant: Thermo Fisher Scientific

385031000
ACRO385031000EA 60.4 EUR
ACRO385031000
Graphit, Pulver
Graphit
Formel: C
Molecular Weight: 12.01 g/mol
Schmelzpunkt: 3827
Lagertemperatur: Raumtemperatur
MDL: MFCD00144065
CAS-Nummer: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560

Bestellen

Spezifikation Testergebnisse

Appearance Black Powder
Ash ≤0.8 %
Carbon (C) ≥99.0 %
Sulfur (S) ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR