Zellulose, mikrokristallin ∼ Ø 50 µm

Lieferant: Thermo Fisher Scientific

Synonyme: Zellulose

387240010 387240050 387242500
ACRO387240010EA 185 EUR
ACRO387240010 ACRO387240050 ACRO387242500
Zellulose, mikrokristallin ∼ Ø 50 µm
Zellulose
Formel: (C₁₂H₂₂O₁₁)n
Schmelzpunkt: 260 - 270 °C
Dichte: 1,27 to 1.6 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00081512
CAS-Nummer: 9004-34-6
EINECS: 232-674-9
Merck Index: 13,01977

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1.5 %
Particle size ≥50 % (> 32 µm)
Residue after ignition ≤0.05 %
pH 5 to 7
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR