Benzyl-(dimethoxyphosphoryl)acetat 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Dimethylphosphonoacetic Acid Benzyl Ester, Dimethyl (Cbz-methyl)phosphonate, Benzyl 2-(dimethoxyphosphoryl)acetate, Dimethyl (Benzyloxycarbonylmethyl)phosphonate

387370050 387370250
ACRO387370050EA 115 EUR
ACRO387370050 ACRO387370250
Benzyl-(dimethoxyphosphoryl)acetat 98%
Benzyl dimethylphosphonoacetate

Appearance: Clear colorless to light brown Liquid

Formel: C₁₁H₁₅O₅P
Siedepunkt: 148…150 °C (,1 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD01872507
CAS-Nummer: 57443-18-2

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5050 to 1.5070 (20°C, 589 nm)
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR