Bis(trifluoromethanesulphonyl)imide 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
lithium bis(trifluoromethanesulphonimide), 1,1,1-Trifluoro-N-((trifluoromethyl)sulphonyl)methanesulphonamide, N,N-Bis(trifluoromethylsulphonyl)amine
387600010
387600050
387600250
ACRO387600010EA
48.8
EUR
ACRO387600010
ACRO387600050
ACRO387600250
Bis(trifluoromethanesulphonyl)imide 99%
Bis(trifluoromethanesulphonyl)imide
Formel:
C₂HF₆NO₄S₂ Siedepunkt: 90…91 °C (760 mmHg) Schmelzpunkt: 52…56 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00214154 CAS-Nummer: 82113-65-3 UN: 3261 ADR: 8,I |
Spezifikation Testergebnisse
Appearance | Colorless to brown Crystals |
Infrared spectrum | Conforms |
Melting point | 50 to 56 °C |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...