Tetrahydro-2H-pyran-2-ol 90% cyclized form of 5-Hydroxypentanal

Lieferant: Thermo Scientific
Achtung

Synonyme: ~1-Valerolactol, 2-Hydroxytetrahydropyran

387900250
ACRO387900250EA 173 EUR
ACRO387900250
Tetrahydro-2H-pyran-2-ol 90% cyclized form of 5-Hydroxypentanal
Tetrahydro-2H-pyran-2-ol

CAS No.: 694-54-2

Formel: C₅H₁₀O₂
Molecular Weight: 102,133 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02683093
CAS-Nummer: 694-54-2

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR