D(+)-Glucose, wasserfrei Ph. Eur., USP, BP

Lieferant: Thermo Fisher Scientific

388190010 388190050 388190250
ACRO388190010EA 37 EUR
ACRO388190010 ACRO388190050 ACRO388190250
D(+)-Glucose, wasserfrei Ph. Eur., USP, BP
D(+)-Glucose
Formel: C₆H₁₂O₆
Schmelzpunkt: 146 °C
Dichte: 1,54 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00063684
CAS-Nummer: 50-99-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white Crystalline powder
Assay 97.5 to 102.0 % (Anhydrous substance)
Water ≤1.0 % (K.F.)
Loss on drying ≤0.5 % (1 g, 105°C, 16 h)
Sulfated ash ≤0.1 % (1 g, 600°C)
Appearance of solution (EP) Passes test
Identification Passes test (EP, USP)
Related substances ≤0.5 % (total impurities)
Specific optical rotation +52.5° to +53.3° (20°C, 589 nm) (C=10,H2O/dilute NH3) (Anhydrous substance)
Acidity (USP) Passes test
Color of solution (USP) Passes test
Chloride (Cl) ≤0.018 %
Sulfate (SO4) ≤0.025 %
Arsenic (As) ≤1 µg/g
Soluble starch, sulfite (USP) Passes test
Dextrin (EP, USP) Passes test
Conductivity ≤20 µS/cm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR