Gold(I)chlorid 99+%

Lieferant: Thermo Fisher Scientific

388330010 388330050
ACRO388330010EA 272 EUR
ACRO388330010 ACRO388330050
Gold(I)chlorid 99+%
Gold(I)chlorid
Formel: AuCl
Molecular Weight: 232.42 g/mol
Schmelzpunkt: 289 °C
MDL: MFCD00046175
CAS-Nummer: 10294-29-8
EINECS: 233-655-8
UN: 3260
ADR: 8 II
Merck Index: 12,04528

Bestellen

Spezifikation Testergebnisse

Appearance Yellow-green Powder
Elemental Analysis ≥99.0 % (ex Au)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR