Ammonium trifluoromethanesulphonate 99%

Lieferant: Thermo Fisher Scientific
Achtung

388730050 388730250 388730500
ACRO388730050EA 66.4 EUR
ACRO388730050 ACRO388730250 ACRO388730500
Ammonium trifluoromethanesulphonate 99%
Ammonium trifluoromethanesulphonate
Formel: CH₄F₃NO₃S
Schmelzpunkt: 225 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00075332
CAS-Nummer: 38542-94-8

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Elemental Analysis ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR