Ammonium tetrathiomolybdate 99.95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Ammonium tetrathiomolybdate(VI)

389530010 389530100 389530500 389532500
ACRO389530010EA 41.8 EUR
ACRO389530010 ACRO389530100 ACRO389530500 ACRO389532500
Ammonium tetrathiomolybdate 99.95%
Ammonium tetrathiomolybdate
Formel: H₁₀MoN₂S₄
Schmelzpunkt: 300 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00136013
CAS-Nummer: 15060-55-6

Bestellen

Spezifikation Testergebnisse

Appearance Brown to black Powder or crystals
Total trace metal impurities ≤500 ppm
Molybdenum (Mo) 35.5 to 38.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR