Rhodium(II) trifluoroacetate dimer

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Trifluoroacetic acid rhodium(II) salt dimer

389700010 389702500
ACRO389700010EA 520 EUR
ACRO389700010 ACRO389702500
Rhodium(II) trifluoroacetate dimer
Rhodium(II) trifluoroacetate dimer
Formel: C₈F₁₂O₈Rh₂
Molecular Weight: 657.87 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00209611
CAS-Nummer: 31126-95-1

Bestellen

Spezifikation Testergebnisse

Appearance Green Crystalline powder
Infrared spectrum Conforms
Rhodium (Rh) 27.5 to 35.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR