Calciumhydrogenphosphat Dihydrat 98+%, reinst

Lieferant: Thermo Fisher Scientific
Achtung

389830010 389830050
ACRO389830010EA 43.7 EUR
ACRO389830010 ACRO389830050
Calciumhydrogenphosphat Dihydrat 98+%, reinst
Calciumhydrogenphosphat Dihydrat
Formel: HCaO₄P·2H₂O
Schmelzpunkt: 36 °C
Dichte: 2,44 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149621
CAS-Nummer: 7789-77-7

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white Powder
Titration Complexometric 98.0 to 105.0 % (on dried substance)
Identification Phosphate: positive
Loss on ignition 24.5 % to 26.5 % (800°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR