Palladium(II)nitrat Hydrat

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Palladiumdinitrat Hydrat

389910010 389910050 389910250
ACRO389910010EA 152 EUR
ACRO389910010 ACRO389910050 ACRO389910250
Palladium(II)nitrat Hydrat
Palladium(II)nitrat Hydrat

CAS No.:

Formel: N₂O₆Pd·xH₂O
Molecular Weight: 230.43 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149818
CAS-Nummer: 207596-32-5
UN: 3085
ADR: 5.1 II

Bestellen

Spezifikation Testergebnisse

Appearance Brown to brown-black Crystals
Palladium (Pd) 39 to 46 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR