Eisen(III)phosphat Hydrat, reinst
Lieferant: Thermo Scientific
Achtung
|
Synonyme:
Eisenphosphat Hydrat, Eisen(III)orthophosphat Hydrat
389955000
ACRO389955000EA
92.3
EUR
ACRO389955000
Eisen(III)phosphat Hydrat, reinst
Eisen(III)phosphat Hydrat
Formel:
FeO₄P·xH₂O Molecular Weight: 150,82 g/mol (anhy) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016096 CAS-Nummer: 51833-68-2 |
Spezifikation Testergebnisse
Appearance | White to yellow to pink to brown Powder |
Titration with Na2S2O3 | 26.0 to 32.0 % (as Fe) |
Fluoride (F) | ≤50 ppm |
Arsenic (As) | ≤3 ppm |
Lead (Pb) | ≤4 ppm |
Loss on ignition | ≤32.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...