Amberlite® FPC3500 Ionenaustauscherharz

Lieferant: Thermo Fisher Scientific

389992500 389990010
ACRO389992500EA 37.1 EUR
ACRO389992500 ACRO389990010
Amberlite® FPC3500 Ionenaustauscherharz
Amberlite® FPC3500
Lagertemperatur: Raumtemperatur
MDL: MFCD08460528
CAS-Nummer: 950182-46-4

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Beads
Particle size ≤5.0 % (> 1.180 mm)
Harmonic mean size 0.450 to 0.650 mm
Uniformity coefficient ≤2.00
Moisture holding capacity 60 to 70 % (H)
Total volume capacity ≥2.60 eq/l (H)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR