(Dimethylamino)acetaldehyddimethylacetal 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2,2-Dimethoxy-N,N-dimethylethanamine, N-(2,2-Dimethoxyethyl)dimethylamine

390910010 390910050
ACRO390910010EA 26.3 EUR
ACRO390910010 ACRO390910050
(Dimethylamino)acetaldehyddimethylacetal 97%
(Dimethylamino)acetaldehyddimethylacetal
Formel: C₆H₁₅NO₂
Molecular Weight: 133.1888 g/mol
Siedepunkt: 64…66 °C (12 mmHg)
Flammpunkt: 32 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010205
CAS-Nummer: 38711-20-5
UN: 2920
ADR: 8 II

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4080 to 1.4120 (20°C, 589 nm)
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR