4-tert-Butylbenzylamin 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
391010010
391010050
ACRO391010010EA
30.5
EUR
ACRO391010010
ACRO391010050
4-tert-Butylbenzylamin 97%
4-tert-Butylbenzylamin
Formel:
C₁₁H₁₇N Flammpunkt: 107 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040754 CAS-Nummer: 39895-55-1 UN: 2735 ADR: 8,III |
Spezifikation Testergebnisse
Appearance | Clear light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5200 to 1.5220 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...