(Triphenylphosphin)gold(I)chlorid 99%

Lieferant: Thermo Fisher Scientific
Achtung

391610010 391610050 391612500
ACRO391610010EA 269 EUR
ACRO391610010 ACRO391610050 ACRO391612500
(Triphenylphosphin)gold(I)chlorid 99%
(Triphenylphosphine)gold(I) chloride
Formel: C₁₈H₁₅AuClP
Lagertemperatur: Kühlschrank
MDL: MFCD00009588
CAS-Nummer: 14243-64-2
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Assay ≥98.5 % (ex Au)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR