Magnesium phosphate tribasic hydrate 98+%, rein

Lieferant: Thermo Fisher Scientific

391942500 391940010
ACRO391942500EA 27.6 EUR
ACRO391942500 ACRO391940010
Magnesium phosphate tribasic hydrate 98+%, rein
Magnesium phosphate tribasic hydrate
Formel: H₂Mg₃O₉P₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00150251
CAS-Nummer: 53408-95-0

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Titration excess HCl 98.0 to 102.0 % (On ignited basis)
Loss on ignition ≤32.0 % (425°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR